missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl Trifluoromethanesulfonate 97.0+%, TCI America™

Supplier:  TCI America T160425G

 View more versions of this product

Catalog No. T160425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,2,2-Trifluoroethyl Trifluoromethanesulfonate
Yellow
C3H2F6O3S
25 g
2,2,2-trifluoroethyltrifluoromethanesulfonate, 2,2,2-trifluoroethyl triflate, methanesulfonic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoromethanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluorometanesulfonic acid, 2,2,2-trifluoroethyltrifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluoromethane sulfonate, trifluoroethyltriflate, trifluoroethyl triflate
C(C(F)(F)F)OS(=O)(=O)C(F)(F)F
232.096
232.10
Liquid
6226-25-1
91°C
MFCD00671579
2920
RTMMSCJWQYWMNK-UHFFFAOYSA-N
2,2,2-trifluoroethyl trifluoromethanesulfonate
80366
≥97.0% (GC)

Chemical Identifiers

6226-25-1
232.096
RTMMSCJWQYWMNK-UHFFFAOYSA-N
80366
C(C(F)(F)F)OS(=O)(=O)C(F)(F)F
C3H2F6O3S
MFCD00671579
2,2,2-trifluoroethyltrifluoromethanesulfonate, 2,2,2-trifluoroethyl triflate, methanesulfonic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoromethanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluorometanesulfonic acid, 2,2,2-trifluoroethyltrifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluoromethane sulfonate, trifluoroethyltriflate, trifluoroethyl triflate
2,2,2-trifluoroethyl trifluoromethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : PB2775000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions