missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl Trifluoromethanesulfonate 97.0+%, TCI America™

Catalog No. p-7136869
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
T16045G 5 g
T160425G 25 g
2 options

Catalog No. T16045G

Supplier: TCI America T16045G

Only null left

Chemical Identifiers

CAS 6226-25-1
Molecular Formula C3H2F6O3S
Molecular Weight (g/mol) 232.096
MDL Number MFCD00671579
InChI Key RTMMSCJWQYWMNK-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyltrifluoromethanesulfonate, 2,2,2-trifluoroethyl triflate, methanesulfonic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoromethanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluorometanesulfonic acid, 2,2,2-trifluoroethyltrifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluoromethane sulfonate, trifluoroethyltriflate, trifluoroethyl triflate
PubChem CID 80366
IUPAC Name 2,2,2-trifluoroethyl trifluoromethanesulfonate
SMILES C(C(F)(F)F)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 91°C
Quantity 5 g
UN Number 2920
Formula Weight 232.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethyl Trifluoromethanesulfonate
RTECSNumber PB2775000
TSCA No