missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. M073825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M073825ML 25 mL
M0738100ML 100 mL
M0738500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M073825ML Supplier TCI America Supplier No. M073825ML

Chemical Identifiers

CAS 352-87-4
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.12
MDL Number MFCD00013576,MFCD00274739
InChI Key QTKPMCIBUROOGY-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl methacrylate, trifluoroethyl methacrylate, methacrylic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylmethacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoroethyl ester, 2-methyl-acrylic acid 2,2,2-trifluoro-ethyl ester, 2-propenoic acid, 2-methyl-, trifluoroethyl ester, 2-methyl-2-propenoic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl m, acmc-1af8z
PubChem CID 9608
IUPAC Name 2,2,2-trifluoroethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(F)(F)F

Specifications

Color Colorless
Boiling Point 102°C
Quantity 25 mL
UN Number 3272
Formula Weight 168.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1031
TSCA Yes