accessibility menu, dialog, popup

UserName

CH3H2COOFFF

CAS RN 352-87-4

IUPAC Name: 2,2,2-trifluoroethyl 2-methylprop-2-enoate
Synonyms: trifluoroethyl methacrylate
Molecular Weight (g/mol): 168.12
Molecular Formula: C6H7F3O2
InChi Key: QTKPMCIBUROOGY-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC(F)(F)F
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)