missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Trifluoroethanol 99.0+%, TCI America™
Supplier: TCI America T0435100G
Specifications
| 2,2,2-Trifluoroethanol | |
| -44°C | |
| 74°C | |
| MFCD00004672 | |
| 1986 | |
| RHQDFWAXVIIEBN-UHFFFAOYSA-N | |
| 2,2,2-trifluoroethan-1-ol | |
| 6409 | |
| 100.04 | |
| Liquid |
| 75-89-8 | |
| Colorless | |
| C2H3F3O | |
| 100 g | |
| trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh | |
| OCC(F)(F)F | |
| 100.04 | |
| CHEBI:42330 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 75-89-8 | |
| 100.04 | |
| RHQDFWAXVIIEBN-UHFFFAOYSA-N | |
| 6409 | |
| 2,2,2-trifluoroethan-1-ol |
| C2H3F3O | |
| MFCD00004672 | |
| trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh | |
| CHEBI:42330 | |
| OCC(F)(F)F |
Safety and Handling
EINECSNumber : (9)-0905
RTECSNumber : KM5250000
TSCA : Yes