accessibility menu, dialog, popup

UserName

HOFFF

CAS RN 75-89-8

IUPAC Name: 2,2,2-trifluoroethan-1-ol
Synonyms: trifluoroethanol TFE fluorinol 85
Molecular Weight (g/mol): 100.04
Molecular Formula: C2H3F3O
InChi Key: RHQDFWAXVIIEBN-UHFFFAOYSA-N
SMILES: OCC(F)(F)F
Boiling Point 
  • (3)
  • (6)
  • (2)
Percent Purity 
  • (4)
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (3)
  • (6)
  • (3)
  • (6)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)