missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanol 99.0+%, TCI America™

Catalog No. T043525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T043525G 25 g
T0435100G 100 g
T0435500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T043525G Supplier TCI America Supplier No. T043525G

Chemical Identifiers

CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
MDL Number MFCD00004672
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
Synonym trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
PubChem CID 6409
ChEBI CHEBI:42330
IUPAC Name 2,2,2-trifluoroethan-1-ol
SMILES OCC(F)(F)F

Specifications

Melting Point -44°C
Color Colorless
Boiling Point 74°C
Quantity 25 g
UN Number 1986
Formula Weight 100.04
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethanol
EINECSNumber (9)-0905
RTECSNumber KM5250000
TSCA Yes