missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanol, NMR grade, 99.5%

Catalog No. p-7046948
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1689806 5 g
AAL1689822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1689806 Supplier Thermo Scientific Chemicals Supplier No. L1689806
Only null left

CAS: 75-89-8 | C2H3F3O | 100.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
MDL Number MFCD00004672
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
Synonym trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
PubChem CID 6409
ChEBI CHEBI:42330
IUPAC Name 2,2,2-trifluoroethan-1-ol
SMILES OCC(F)(F)F

Specifications

Melting Point -44°C
Density 1.391
Boiling Point 74°C to 75°C
Flash Point 29°C (84°F)
Refractive Index 1.29
Linear Formula CF3CH2OH
Quantity 5 g
UN Number UN1992
Beilstein 1733203
Formula Weight 100.04
Percent Purity 99.5%
Grade NMR
Chemical Name or Material 2,2,2-Trifluoroethanol
Show More Show Less
Hazard Category H226-H301+H331-H318-H360F-H373
Hazard Statement GHS H Statement
H301-H311-H318-H226-H332-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 200-913-6
RTECSNumber KM5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only