missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanol, 99+%, Reagents

Catalog No. 01336906
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336906 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336906 Supplier Labchem Inc Supplier No. C23426701C
Only null left

2,2,2-Trifluoroethanol, 99+%, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
IUPAC Name 2,2,2-trifluoroethan-1-ol
SMILES OCC(F)(F)F

Specifications

Boiling Range 78°C
Color Colorless
Boiling Point 78°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.43
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethanol, 99+%