missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Trifluoroacetophenone 98.0+%, TCI America™
Supplier: TCI America T084825G
Specifications
| 2,2,2-Trifluoroacetophenone | |
| 166°C | |
| MFCD00000420 | |
| 1224 | |
| KZJRKRQSDZGHEC-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-1-phenylethanone | |
| 9905 | |
| ≥98.0% (GC) |
| 434-45-7 | |
| C8H5F3O | |
| 25 g | |
| trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09 | |
| C1=CC=C(C=C1)C(=O)C(F)(F)F | |
| 174.122 | |
| 174.12 | |
| Liquid |
Chemical Identifiers
| 434-45-7 | |
| 174.122 | |
| KZJRKRQSDZGHEC-UHFFFAOYSA-N | |
| 9905 | |
| C1=CC=C(C=C1)C(=O)C(F)(F)F |
| C8H5F3O | |
| MFCD00000420 | |
| trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09 | |
| 2,2,2-trifluoro-1-phenylethanone |
Safety and Handling
TSCA : Yes