missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroacetophenone 98.0+%, TCI America™

Catalog No. p-7136458
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
T08485G 5 g
T084825G 25 g
2 options

Catalog No. T08485G

Supplier: TCI America T08485G

Only null left

Chemical Identifiers

CAS 434-45-7
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.122
MDL Number MFCD00000420
InChI Key KZJRKRQSDZGHEC-UHFFFAOYSA-N
Synonym trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09
PubChem CID 9905
IUPAC Name 2,2,2-trifluoro-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(F)(F)F

Specifications

Boiling Point 166°C
Quantity 5 g
UN Number 1224
Formula Weight 174.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroacetophenone