missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroacetamide, 97%

Catalog No. p-3211461
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC385220250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC385220250 Supplier Thermo Scientific Chemicals Supplier No. 385220250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 354-38-1 | C2H2F3NO | 113.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 354-38-1
Molecular Formula C2H2F3NO
Molecular Weight (g/mol) 113.04
MDL Number MFCD00008008
InChI Key NRKYWOKHZRQRJR-UHFFFAOYSA-N
Synonym trifluoroacetamide, acetamide, 2,2,2-trifluoro, unii-36mu16byfk, n-trifluoroacetamide, 2,2,2-trifluoro-acetamide, 36mu16byfk, trifluroacetamide, trifluoro-acetamide, snylgqhjptadtaetp@, pubchem12622
PubChem CID 67717
IUPAC Name 2,2,2-trifluoroacetamide
SMILES NC(=O)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 59°C to 70°C
Color White to Yellow or Beige
Boiling Point 162°C to 164°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CF3CONH2
Quantity 25 g
Beilstein 02, II, 186
Solubility Information Solubility in water: 460g/l (20°C). Other solubilities: soluble in alcohol,ether,partially soluble in chloroform
Formula Weight 113.04
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 2, 2-Trifluoroacetamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-559-9

RUO – Research Use Only