missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide, MSTFA, EMD-Millipore

Catalog No. m1118050005
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1118050005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1118050005 Supplier Technidata Supplier No. 1.11805.0005

Chemical Identifiers

CAS 24589-78-4
Molecular Formula C6H12F3NOSi
Molecular Weight (g/mol) 199.25
MDL Number MFCD00000411
InChI Key MSPCIZMDDUQPGJ-UHFFFAOYSA-N
Synonym MSTFA
PubChem CID 32510
ChEBI CHEBI:85064
IUPAC Name 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide
SMILES CN(C(=O)C(F)(F)F)[Si](C)(C)C

Specifications

Packaging Glass
Density Density: 1.085 g/cm3
Quantity 5 mL
Formula Weight 199.25
Chemical Name or Material 2, 2, 2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide
Recommended Storage Store without restrictions