missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Trifluoro-3'-nitroacetophenone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 56°C |
| Boiling Point | 113°C |
| Quantity | 1 g |
| Formula Weight | 219.12 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2,2-Trifluoro-3′-nitroacetophenone |
Safety and Handling
| TSCA | No |