accessibility menu, dialog, popup

UserName

CAS RN 657-15-8

ONOOFFF

CAS RN 657-15-8

IUPAC Name: 2,2,2-trifluoro-1-(3-nitrophenyl)ethan-1-one
Synonyms: 2,2,2-trifluoro-1-(3-nitrophenyl)ethanone
Molecular Weight (g/mol): 219.12
Molecular Formula: C8H4F3NO3
InChi Key: QADCNGZPRUSTJL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results