missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethyl Sulfamate 98.0+%, TCI America™

Catalog No. T26465G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26465G Supplier TCI America Supplier No. T26465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69226-51-3
Molecular Formula C2H4Cl3NO3S
Molecular Weight (g/mol) 228.468
MDL Number MFCD01683146
InChI Key VOKONKGVTXWZJI-UHFFFAOYSA-N
Synonym Sulfamic Acid 2,2,2-Trichloroethyl Ester, 2,2,2-Trichloroethoxysulfonamide
PubChem CID 3052707
IUPAC Name 2,2,2-trichloroethyl sulfamate
SMILES C(C(Cl)(Cl)Cl)OS(=O)(=O)N

Specifications

Melting Point 56°C
Color White
Quantity 5 g
Formula Weight 228.47
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,2-Trichloroethyl Sulfamate
RTECSNumber WO6200000
TSCA No