CAS RN 69226-51-3
IUPAC Name:
2,2,2-trichloroethyl sulfamate
Synonyms:
2,2,2-trichloroethyl sulfamate
Molecular Weight (g/mol):
228.47
Molecular Formula:
C2H4Cl3NO3S
InChi Key:
VOKONKGVTXWZJI-UHFFFAOYSA-N
SMILES:
NS(=O)(=O)OCC(Cl)(Cl)Cl
1
–
1
of
1
results
1
2,2,2-Trichloroethyl Sulfamate 98.0+%, TCI America™
CAS: 69226-51-3 Molecular Formula: C2H4Cl3NO3S Molecular Weight (g/mol): 228.468 MDL Number: MFCD01683146 InChI Key: VOKONKGVTXWZJI-UHFFFAOYSA-N Synonym: Sulfamic Acid 2,2,2-Trichloroethyl Ester, 2,2,2-Trichloroethoxysulfonamide PubChem CID: 3052707 IUPAC Name: 2,2,2-trichloroethyl sulfamate SMILES: C(C(Cl)(Cl)Cl)OS(=O)(=O)N