missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethyl Chloroformate 98.0+%, TCI America™

Catalog No. p-7124613
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C079525G 25 g
C0795250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C079525G Supplier TCI America Supplier No. C079525G
Only null left

Chemical Identifiers

CAS 17341-93-4
Molecular Formula C3H2Cl4O2
Molecular Weight (g/mol) 211.847
MDL Number MFCD00000810
InChI Key LJCZNYWLQZZIOS-UHFFFAOYSA-N
Synonym 2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc
PubChem CID 87063
IUPAC Name 2,2,2-trichloroethyl carbonochloridate
SMILES C(C(Cl)(Cl)Cl)OC(=O)Cl

Specifications

Color Colorless
Boiling Point 172°C
Quantity 25 g
UN Number 3277
Formula Weight 211.85
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trichloroethyl Chloroformate
EINECSNumber (2)-3319
TSCA Yes