missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethyl chloroformate, 97%

CAS: 17341-93-4 | C3H2Cl4O2 | 211.847 g/mol

Supplier:  Thermo Scientific Chemicals L0687522

 View more versions of this product

Catalog No. AAL0687522


Only null left
Explore available promotions

Description

Description

2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.

Solubility
Soluble in Ether, Benzene, Chloroform, Ethanol. Insoluble in water.

Notes
Moisture Sensitive. Desiccate at 4°C. Store under inert gas. Incompatible materials are Strong oxidizing agents.
Specifications

Specifications

2,2,2-Trichloroethyl chloroformate
0°C to 2°C
171°C to 172°C
C3H2Cl4O2
MFCD00000810
UN3277
Moisture sensitive
Soluble in Ether,Benzene,Chloroform,Ethanol. Insoluble in water.
C(C(Cl)(Cl)Cl)OC(=O)Cl
211.847
211.86
17341-93-4
1.539
>100°C (212°F)
1.47
100 g
970619
2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc
LJCZNYWLQZZIOS-UHFFFAOYSA-N
2,2,2-trichloroethyl carbonochloridate
87063
97%

Chemical Identifiers

17341-93-4
211.847
LJCZNYWLQZZIOS-UHFFFAOYSA-N
87063
C(C(Cl)(Cl)Cl)OC(=O)Cl
C3H2Cl4O2
MFCD00000810
2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc
2,2,2-trichloroethyl carbonochloridate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H331-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, N.O.S.

EINECSNumber : 241-363-7

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only