missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethyl chloroformate, 97%

Catalog No. AAL0687514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0687514 25 g
AAL0687522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0687514 Supplier Thermo Scientific Chemicals Supplier No. L0687514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17341-93-4 | C3H2Cl4O2 | 211.847 g/mol

2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.

Solubility
Soluble in Ether, Benzene, Chloroform, Ethanol. Insoluble in water.

Notes
Moisture Sensitive. Desiccate at 4°C. Store under inert gas. Incompatible materials are Strong oxidizing agents.

Chemical Identifiers

CAS 17341-93-4
Molecular Formula C3H2Cl4O2
Molecular Weight (g/mol) 211.847
MDL Number MFCD00000810
InChI Key LJCZNYWLQZZIOS-UHFFFAOYSA-N
Synonym 2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc
PubChem CID 87063
IUPAC Name 2,2,2-trichloroethyl carbonochloridate
SMILES C(C(Cl)(Cl)Cl)OC(=O)Cl

Specifications

Melting Point 0°C to 2°C
Density 1.539
Boiling Point 171°C to 172°C
Flash Point >100°C (212°F)
Refractive Index 1.47
Quantity 25 g
UN Number UN3277
Beilstein 970619
Sensitivity Moisture sensitive
Solubility Information Soluble in Ether,Benzene,Chloroform,Ethanol. Insoluble in water.
Formula Weight 211.86
Percent Purity 97%
Chemical Name or Material 2,2,2-Trichloroethyl chloroformate
Show More Show Less
Hazard Category H302-H314-H331-H335
Hazard Statement GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, N.O.S.
EINECSNumber 241-363-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only