missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethanol 98.0+%, TCI America™

Supplier:  TCI America T0622500G

 View more versions of this product

Catalog No. T0622500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,2,2-Trichloroethanol
18°C
151°C
MFCD00004677
1760
KPWDGTGXUYRARH-UHFFFAOYSA-N
2,2,2-trichloroethanol
8259
149.40
Liquid
115-20-8
Yellow
C2H3Cl3O
500 g
trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol
C(C(Cl)(Cl)Cl)O
149.395
CHEBI:28094
≥98.0% (GC)

Chemical Identifiers

115-20-8
149.395
KPWDGTGXUYRARH-UHFFFAOYSA-N
8259
2,2,2-trichloroethanol
C2H3Cl3O
MFCD00004677
trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol
CHEBI:28094
C(C(Cl)(Cl)Cl)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2002

RTECSNumber : KM3850000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions