missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(2-Methoxyethoxy)ethoxy]ethyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. p-7133561
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M3094100G 100 g
M3094500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M3094100G Supplier TCI America Supplier No. M3094100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 48067-72-7
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.249
MDL Number MFCD30470140
InChI Key NMLCFUMBGQIRJX-UHFFFAOYSA-N
Synonym Acrylic Acid 2-[2-(2-Methoxyethoxy)ethoxy]ethyl Ester
PubChem CID 3016467
IUPAC Name 2-[2-(2-methoxyethoxy)ethoxy]ethyl prop-2-enoate
SMILES COCCOCCOCCOC(=O)C=C

Specifications

Color Yellow
Quantity 100 g
Formula Weight 218.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-[2-(2-Methoxyethoxy)ethoxy]ethyl Acrylate (stabilized with MEHQ)