accessibility menu, dialog, popup

UserName

H3COOOOOCH2

CAS RN 48067-72-7

IUPAC Name: 2-[2-(2-methoxyethoxy)ethoxy]ethyl prop-2-enoate
Synonyms: 2-[2-(2-methoxyethoxy)ethoxy]ethyl prop-2-enoate
Molecular Weight (g/mol): 218.25
Molecular Formula: C10H18O5
InChi Key: NMLCFUMBGQIRJX-UHFFFAOYSA-N
SMILES: COCCOCCOCCOC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results