missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[2-(2-Aminoethoxy)ethoxy]acetic Acid 96.0+%, TCI America™
$210.75 - $634.13
Chemical Identifiers
| CAS | 134978-97-5 |
|---|---|
| Molecular Formula | C6H13NO4 |
| Molecular Weight (g/mol) | 163.17 |
| MDL Number | MFCD13185897 |
| InChI Key | RUVRGYVESPRHSZ-UHFFFAOYSA-N |
| Synonym | 8-Amino-3,6-dioxaoctanoic Acid, Amino-PEG2-acetic Acid |
| PubChem CID | 362706 |
| IUPAC Name | 2-[2-(2-aminoethoxy)ethoxy]acetic acid |
| SMILES | NCCOCCOCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A3120250MG
|
TCI America
A3120250MG |
250 mg |
Each for $210.75
|
|
|||||
|
A31201G
|
TCI America
A31201G |
1 g |
Each for $634.13
|
|
|||||
Chemical Identifiers
| 134978-97-5 | |
| 163.17 | |
| RUVRGYVESPRHSZ-UHFFFAOYSA-N | |
| 362706 | |
| NCCOCCOCC(O)=O |
| C6H13NO4 | |
| MFCD13185897 | |
| 8-Amino-3,6-dioxaoctanoic Acid, Amino-PEG2-acetic Acid | |
| 2-[2-(2-aminoethoxy)ethoxy]acetic acid |
Specifications
| 134978-97-5 | |
| MFCD13185897 | |
| RUVRGYVESPRHSZ-UHFFFAOYSA-N | |
| 2-[2-(2-aminoethoxy)ethoxy]acetic acid | |
| 163.17 | |
| 163.17 | |
| Crystalline Powder |
| C6H13NO4 | |
| 8-Amino-3,6-dioxaoctanoic Acid, Amino-PEG2-acetic Acid | |
| NCCOCCOCC(O)=O | |
| 250 mg | |
| 362706 | |
| ≥96.0% (T) | |
| 2-[2-(2-Aminoethoxy)ethoxy]acetic Acid |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)