CAS RN 134978-97-5
CAS RN 134978-97-5
IUPAC Name:
2-[2-(2-aminoethoxy)ethoxy]acetic acid
Synonyms:
[2-(2-aminoethoxy)ethoxy]acetic acid
Molecular Weight (g/mol):
163.17
Molecular Formula:
C6H13NO4
InChi Key:
RUVRGYVESPRHSZ-UHFFFAOYSA-N
SMILES:
NCCOCCOCC(O)=O
1
–
1
of
1
results
1
2-[2-(2-Aminoethoxy)ethoxy]acetic Acid 96.0+%, TCI America™
CAS: 134978-97-5 Molecular Formula: C6H13NO4 Molecular Weight (g/mol): 163.17 MDL Number: MFCD13185897 InChI Key: RUVRGYVESPRHSZ-UHFFFAOYSA-N Synonym: 8-Amino-3,6-dioxaoctanoic Acid, Amino-PEG2-acetic Acid PubChem CID: 362706 IUPAC Name: 2-[2-(2-aminoethoxy)ethoxy]acetic acid SMILES: NCCOCCOCC(O)=O