missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane) 98.0+%, TCI America™
Supplier: TCI America O046925G
Specifications
| 2,2′-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane) | |
| Yellow | |
| C10H10F8O2 | |
| 25 g | |
| FC(F)(CC1CO1)C(F)(F)C(F)(F)C(F)(F)CC1CO1 | |
| 314.18 | |
| 314.18 | |
| Liquid |
| 791-22-0 | |
| 112°C | |
| MFCD01320774 | |
| KVSHGEMJMXSNTB-UHFFFAOYNA-N | |
| 2-[2,2,3,3,4,4,5,5-octafluoro-6-(oxiran-2-yl)hexyl]oxirane | |
| 2736025 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 791-22-0 | |
| 314.18 | |
| KVSHGEMJMXSNTB-UHFFFAOYNA-N | |
| 2-[2,2,3,3,4,4,5,5-octafluoro-6-(oxiran-2-yl)hexyl]oxirane |
| C10H10F8O2 | |
| MFCD01320774 | |
| 2736025 | |
| FC(F)(CC1CO1)C(F)(F)C(F)(F)C(F)(F)CC1CO1 |
Safety and Handling
TSCA : No