accessibility menu, dialog, popup

UserName

OOFFFFFFFF

CAS RN 791-22-0

IUPAC Name: 2-[2,2,3,3,4,4,5,5-octafluoro-6-(oxiran-2-yl)hexyl]oxirane
Synonyms: 2-[2,2,3,3,4,4,5,5-octafluoro-6-(oxiran-2-yl)hexyl]oxirane
Molecular Weight (g/mol): 314.18
Molecular Formula: C10H10F8O2
InChi Key: KVSHGEMJMXSNTB-UHFFFAOYNA-N
SMILES: FC(F)(CC1CO1)C(F)(F)C(F)(F)C(F)(F)CC1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results