missing translation for 'onlineSavingsMsg'
Learn More

2,2'-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane) 98.0+%, TCI America™

Catalog No. O04695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04695G 5 g
O046925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04695G Supplier TCI America Supplier No. O04695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 791-22-0
Molecular Formula C10H10F8O2
Molecular Weight (g/mol) 314.18
MDL Number MFCD01320774
InChI Key KVSHGEMJMXSNTB-UHFFFAOYNA-N
PubChem CID 2736025
IUPAC Name 2-[2,2,3,3,4,4,5,5-octafluoro-6-(oxiran-2-yl)hexyl]oxirane
SMILES FC(F)(CC1CO1)C(F)(F)C(F)(F)C(F)(F)CC1CO1

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 g
Formula Weight 314.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2′-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane)