missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1H-Pyrazol-3-yl)phenol 98.0+%, TCI America™
$121.16 - $430.60
Chemical Identifiers
| CAS | 34810-67-8 |
|---|---|
| Molecular Formula | C9H8N2O |
| Molecular Weight (g/mol) | 160.176 |
| MDL Number | MFCD03093876 |
| InChI Key | SUWASPSYOCZZQV-UHFFFAOYSA-N |
| Synonym | 3-(2-Hydroxyphenyl)pyrazole |
| PubChem CID | 6743769 |
| IUPAC Name | 6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one |
| SMILES | C1=CC(=C2C=CNN2)C(=O)C=C1 |
Chemical Identifiers
| 34810-67-8 | |
| 160.176 | |
| SUWASPSYOCZZQV-UHFFFAOYSA-N | |
| 6743769 | |
| C1=CC(=C2C=CNN2)C(=O)C=C1 |
| C9H8N2O | |
| MFCD03093876 | |
| 3-(2-Hydroxyphenyl)pyrazole | |
| 6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one |
Specifications
| 34810-67-8 | |
| White-Yellow | |
| MFCD03093876 | |
| 3-(2-Hydroxyphenyl)pyrazole | |
| C1=CC(=C2C=CNN2)C(=O)C=C1 | |
| 160.176 | |
| 160.18 | |
| Crystalline Powder |
| 97°C | |
| C9H8N2O | |
| 1 g | |
| SUWASPSYOCZZQV-UHFFFAOYSA-N | |
| 6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one | |
| 6743769 | |
| ≥98.0% (GC,T) | |
| 2-(1H-Pyrazol-3-yl)phenol |
Safety and Handling
TSCA : No