accessibility menu, dialog, popup

UserName

CAS RN 34810-67-8

OHNHN(Z)

CAS RN 34810-67-8

IUPAC Name: 6-[(3Z)-2,3-dihydro-1H-pyrazol-3-ylidene]cyclohexa-2,4-dien-1-one
Synonyms: 6-[(3Z)-1,2-dihydropyrazol-3-ylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: SUWASPSYOCZZQV-FPLPWBNLSA-N
SMILES: O=C1C=CC=C\C1=C1\NNC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results