missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1H-Pyrazol-1-yl)pyridine, Thermo Scientific™
$107.22 - $360.40
Chemical Identifiers
| CAS | 25700-11-2 |
|---|---|
| Molecular Formula | C8H7N3 |
| Molecular Weight (g/mol) | 145.17 |
| MDL Number | 09033534 |
| InChI Key | XXTPHXNBKRVYJI-UHFFFAOYSA-N |
| IUPAC Name | 2-(1H-pyrazol-1-yl)pyridine |
| SMILES | C1=CN(N=C1)C1=CC=CC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC469560010
|
Thermo Scientific Chemicals
469560010 |
1 g |
Each for $107.22
|
|
|||||
|
AC469560050
|
Thermo Scientific Chemicals
469560050 |
5 g |
Each for $360.40
|
|
|||||
Chemical Identifiers
| 25700-11-2 | |
| 145.17 | |
| XXTPHXNBKRVYJI-UHFFFAOYSA-N | |
| C1=CN(N=C1)C1=CC=CC=N1 |
| C8H7N3 | |
| 09033534 | |
| 2-(1H-pyrazol-1-yl)pyridine |
Specifications
| PYRAZOLYLPYRIDINE | |
| 35°C to 45°C | |
| Conforms | |
| C8H7N3 | |
| 1 g | |
| XXTPHXNBKRVYJI-UHFFFAOYSA-N | |
| 2-(1H-pyrazol-1-yl)pyridine | |
| Crystalline Powder and/or Chunks |
| 25700-11-2 | |
| White to almost white | |
| Glass Bottle | |
| 09033534 | |
| Soluble in methanol | |
| C1=CN(N=C1)C1=CC=CC=N1 | |
| 145.17 | |
| 2-(1H-Pyrazol-1-yl)pyridine |
Safety and Handling
Exclamation mark
Recommended Storage : Normal conditions