accessibility menu, dialog, popup

UserName

NNN

CAS RN 25700-11-2

IUPAC Name: 2-(1H-pyrazol-1-yl)pyridine
Synonyms: 2-(pyrazol-1-yl)pyridine
Molecular Weight (g/mol): 145.17
Molecular Formula: C8H7N3
InChi Key: XXTPHXNBKRVYJI-UHFFFAOYSA-N
SMILES: C1=CN(N=C1)C1=CC=CC=N1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)