missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Pyrazol-1-yl)pyridine, Thermo Scientific™

Catalog No. AC469560050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469560050 5 g
AC469560010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC469560050 Supplier Thermo Scientific Chemicals Supplier No. 469560050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25700-11-2
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number 09033534
InChI Key XXTPHXNBKRVYJI-UHFFFAOYSA-N
IUPAC Name 2-(1H-pyrazol-1-yl)pyridine
SMILES C1=CN(N=C1)C1=CC=CC=N1

Specifications

Alpha Vector PYRAZOLYLPYRIDINE
Melting Point 35°C to 45°C
Color White to almost white
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 5 g
Solubility Information Soluble in methanol
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-(1H-Pyrazol-1-yl)pyridine
Hazard Category Exclamation mark
Recommended Storage Normal conditions