missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1H-Pyrazol-1-yl)pyridine, Thermo Scientific™
Specifications
Specifications
| Alpha Vector | PYRAZOLYLPYRIDINE |
| Melting Point | 35°C to 45°C |
| Color | White to almost white |
| Infrared Spectrum | Conforms |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Solubility Information | Soluble in methanol |
| Physical Form | Crystalline Powder and/or Chunks |
| Chemical Name or Material | 2-(1H-Pyrazol-1-yl)pyridine |
Safety and Handling
| Hazard Category | Exclamation mark |
| Recommended Storage | Normal conditions |