missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Imidazol-1-yl)aniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC32714EA

Quantity 10g

 View more versions of this product

Catalog No. CC32714EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

26286-54-4
159.192
HVECTIQVQPUSEX-UHFFFAOYSA-N
320166
C1=CC=C(C(=C1)N)N2C=CN=C2
C9H9N3
MFCD03274531
2-1h-imidazol-1-yl aniline, 2-1-imidazolyl aniline, 2-imidazol-1-yl-phenylamine, 2-imidazol-1-yl aniline, benzenamine,2-1h-imidazol-1-yl, 2-imidazolylphenylamine, 2-1h-imidazol-1-yl benzenamine, pubchem10103, n-o-aminophenyl imidazole, 2-imidazol-1-ylphenylamine
2-imidazol-1-ylaniline

Specifications

2-(1H-Imidazol-1-yl)aniline
96.5
97%
10g
2-1h-imidazol-1-yl aniline, 2-1-imidazolyl aniline, 2-imidazol-1-yl-phenylamine, 2-imidazol-1-yl aniline, benzenamine,2-1h-imidazol-1-yl, 2-imidazolylphenylamine, 2-1h-imidazol-1-yl benzenamine, pubchem10103, n-o-aminophenyl imidazole, 2-imidazol-1-ylphenylamine
C1=CC=C(C(=C1)N)N2C=CN=C2
159.192
159.19
26286-54-4
100.0
C9H9N3
MFCD03274531
HVECTIQVQPUSEX-UHFFFAOYSA-N
2-imidazol-1-ylaniline
320166
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents