accessibility menu, dialog, popup

UserName

CAS RN 26286-54-4

IUPAC Name: 2-(1H-imidazol-1-yl)aniline
Synonyms: 2-(imidazol-1-yl)aniline
Molecular Weight (g/mol): 159.19
Molecular Formula: C9H9N3
InChi Key: HVECTIQVQPUSEX-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1N1C=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)