missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Imidazol-1-yl)aniline, 97%, Thermo Scientific™

Catalog No. CC32714CB
Change view
Click to view available options
Quantity:
250mg
1g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC32714CB 250mg
CC32714DA 1g
CC32714EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC32714CB Supplier Thermo Scientific Chemicals Supplier No. CC32714CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26286-54-4
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
MDL Number MFCD03274531
InChI Key HVECTIQVQPUSEX-UHFFFAOYSA-N
Synonym 2-1h-imidazol-1-yl aniline, 2-1-imidazolyl aniline, 2-imidazol-1-yl-phenylamine, 2-imidazol-1-yl aniline, benzenamine,2-1h-imidazol-1-yl, 2-imidazolylphenylamine, 2-1h-imidazol-1-yl benzenamine, pubchem10103, n-o-aminophenyl imidazole, 2-imidazol-1-ylphenylamine
PubChem CID 320166
IUPAC Name 2-imidazol-1-ylaniline
SMILES C1=CC=C(C(=C1)N)N2C=CN=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 2-(1H-Imidazol-1-yl)aniline
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.