missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-1,2,4-Triazol-3-yl)pyridine 98.0+%, TCI America™

Catalog No. P2100200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2100200MG 200 mg
P21001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2100200MG Supplier TCI America Supplier No. P2100200MG

Chemical Identifiers

CAS 23195-62-2
Molecular Formula C7H6N4
Molecular Weight (g/mol) 146.153
MDL Number MFCD13188589
InChI Key PVMLJDLTWMGZAH-UHFFFAOYSA-N
Synonym 3-(2-Pyridyl)-1,2,4-triazole
PubChem CID 10898842
IUPAC Name 2-(1H-1,2,4-triazol-5-yl)pyridine
SMILES C1=CC=NC(=C1)C2=NC=NN2

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 200 mg
Formula Weight 146.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1H-1,2,4-Triazol-3-yl)pyridine