accessibility menu, dialog, popup

UserName

NNNHN

CAS RN 23195-62-2

IUPAC Name: 2-(1H-1,2,4-triazol-5-yl)pyridine
Synonyms: 2-(2H-1,2,4-triazol-3-yl)pyridine
Molecular Weight (g/mol): 146.15
Molecular Formula: C7H6N4
InChi Key: PVMLJDLTWMGZAH-UHFFFAOYSA-N
SMILES: N1N=CN=C1C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results