missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-1,2,4-Triazol-1-yl)-2',4'-difluoroacetophenone 98.0+%, TCI America™

Supplier:  TCI America T272225G

 View more versions of this product

Catalog No. T272225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(1H-1,2,4-Triazol-1-yl)-2′,4′-difluoroacetophenone
109°C
MFCD02093825
1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethanone, 2,4-difluoro-alpha-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1,2,4-triazol-1-yl ethanone, 2',4'-difluoro-2-1h-1,2,4-triazol-1-yl acetophenone, unii-hxi8r9r915, 2-1h-1,2,4-triazol-1-yl-2',4'-difluoroacetophenone, ethanone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl, 2',4'-difluoro-2-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethan-1-one, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl-ethanone
C1=CC(=C(C=C1F)F)C(=O)CN2C=NC=N2
223.183
223.18
Crystalline Powder
86404-63-9
C10H7F2N3O
25 g
XCHRPVARHBCFMJ-UHFFFAOYSA-N
1-(2,4-difluorophenyl)-2-(1,2,4-triazol-1-yl)ethanone
588080
≥98.0% (GC)

Chemical Identifiers

86404-63-9
223.183
XCHRPVARHBCFMJ-UHFFFAOYSA-N
588080
C1=CC(=C(C=C1F)F)C(=O)CN2C=NC=N2
C10H7F2N3O
MFCD02093825
1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethanone, 2,4-difluoro-alpha-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1,2,4-triazol-1-yl ethanone, 2',4'-difluoro-2-1h-1,2,4-triazol-1-yl acetophenone, unii-hxi8r9r915, 2-1h-1,2,4-triazol-1-yl-2',4'-difluoroacetophenone, ethanone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl, 2',4'-difluoro-2-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethan-1-one, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl-ethanone
1-(2,4-difluorophenyl)-2-(1,2,4-triazol-1-yl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions