missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-1,2,4-Triazol-1-yl)-2',4'-difluoroacetophenone 98.0+%, TCI America™

Catalog No. p-7137373
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27225G 5 g
T272225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27225G Supplier TCI America Supplier No. T27225G
Only null left

Chemical Identifiers

CAS 86404-63-9
Molecular Formula C10H7F2N3O
Molecular Weight (g/mol) 223.183
MDL Number MFCD02093825
InChI Key XCHRPVARHBCFMJ-UHFFFAOYSA-N
Synonym 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethanone, 2,4-difluoro-alpha-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1,2,4-triazol-1-yl ethanone, 2',4'-difluoro-2-1h-1,2,4-triazol-1-yl acetophenone, unii-hxi8r9r915, 2-1h-1,2,4-triazol-1-yl-2',4'-difluoroacetophenone, ethanone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl, 2',4'-difluoro-2-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethan-1-one, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl-ethanone
PubChem CID 588080
IUPAC Name 1-(2,4-difluorophenyl)-2-(1,2,4-triazol-1-yl)ethanone
SMILES C1=CC(=C(C=C1F)F)C(=O)CN2C=NC=N2

Specifications

Melting Point 109°C
Quantity 5 g
Formula Weight 223.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1H-1,2,4-Triazol-1-yl)-2′,4′-difluoroacetophenone