missing translation for 'onlineSavingsMsg'
Learn More

2,12-Dichlorocyclododecanone 98.0+%, TCI America™

Catalog No. D29111G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29111G Supplier TCI America Supplier No. D29111G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 858830-26-9
Molecular Formula C12H20Cl2O
Molecular Weight (g/mol) 251.19
MDL Number MFCD04117917
InChI Key MEHFXKFLCAZPSE-UHFFFAOYNA-N
PubChem CID 44630272
IUPAC Name 2,12-dichlorocyclododecan-1-one
SMILES ClC1CCCCCCCCCC(Cl)C1=O

Specifications

Color White
Quantity 1 g
Formula Weight 251.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,12-Dichlorocyclododecanone
TSCA No
Recommended Storage Refrigerator