accessibility menu, dialog, popup

UserName

OClCl

CAS RN 858830-26-9

IUPAC Name: 2,12-dichlorocyclododecan-1-one
Synonyms: 2,12-dichlorocyclododecan-1-one
Molecular Weight (g/mol): 251.19
Molecular Formula: C12H20Cl2O
InChi Key: MEHFXKFLCAZPSE-UHFFFAOYNA-N
SMILES: ClC1CCCCCCCCCC(Cl)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results