missing translation for 'onlineSavingsMsg'
Learn More

2-[(11bR)-3H-Binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylamine, 97+%, Thermo Scientific Chemicals

$402.49 - $402.49

Chemical Identifiers

CAS 1092064-00-0
Molecular Formula C24H22NP
Molecular Weight (g/mol) 355.421
MDL Number MFCD11045442
InChI Key PXDLCEFIOZLJLW-UHFFFAOYSA-N
Synonym 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethyl amine, 2-11br-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylamine, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethanamine, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethanamine, kanata purity, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylamine, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethyl amine, 2-11bs-3h-binaphtho 2,1-c:1 inverted exclamation marka,2 inverted exclamation marka-e phosphepin-4 5h-yl ethanamine, 2-13-phosphapentacyclo 13.8.0.0 2 , 1 1 .0 3 ,?.0 1 ?, 2 3 tricosa-1 15 ,2 11 ,3,5,7,9,16,18,20,22-decaen-13-yl ethanamine, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanamine
PubChem CID 46177751
SMILES C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CCN)C=CC5=CC=CC=C54
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60423MD
View Documents
Thermo Scientific Chemicals
H60423MD
250 mg
Each for $402.49
Only null left
 
Description

Description

2-[(11bR)-3H-Binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylamine, is used as a application of innovative catalyst, ligand synthesis and application in asymmetric catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-[(11bR)-3H-Binaphtho[2,1-c:1′,2′-e]phosphepin-4(5H)-yl]ethylamine, is used as a application of innovative catalyst, ligand synthesis and application in asymmetric catalysis.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Air, sensitive.
Specifications

Chemical Identifiers

1092064-00-0
355.421
PXDLCEFIOZLJLW-UHFFFAOYSA-N
46177751
C24H22NP
MFCD11045442
2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethyl amine, 2-11br-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylamine, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethanamine, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethanamine, kanata purity, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylamine, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethyl amine, 2-11bs-3h-binaphtho 2,1-c:1 inverted exclamation marka,2 inverted exclamation marka-e phosphepin-4 5h-yl ethanamine, 2-13-phosphapentacyclo 13.8.0.0 2 , 1 1 .0 3 ,?.0 1 ?, 2 3 tricosa-1 15 ,2 11 ,3,5,7,9,16,18,20,22-decaen-13-yl ethanamine, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanamine
C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CCN)C=CC5=CC=CC=C54

Specifications

1092064-00-0
MFCD11045442
Air Sensitive
Soluble in water.
C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CCN)C=CC5=CC=CC=C54
46177751
≥97%
C24H22NP
250 mg
2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethyl amine, 2-11br-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylamine, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethanamine, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethanamine, kanata purity, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylamine, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethyl amine, 2-11bs-3h-binaphtho 2,1-c:1 inverted exclamation marka,2 inverted exclamation marka-e phosphepin-4 5h-yl ethanamine, 2-13-phosphapentacyclo 13.8.0.0 2 , 1 1 .0 3 ,?.0 1 ?, 2 3 tricosa-1 15 ,2 11 ,3,5,7,9,16,18,20,22-decaen-13-yl ethanamine, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanamine
PXDLCEFIOZLJLW-UHFFFAOYSA-N
355.421
355.41
2-[(11bR)-3H-Binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only