accessibility menu, dialog, popup

UserName

CAS RN 1092064-00-0

IUPAC Name: 2-{13-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-yl}ethan-1-amine
Synonyms: 2-{13-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-yl}ethanamine
Molecular Weight (g/mol): 355.42
Molecular Formula: C24H22NP
InChi Key: PXDLCEFIOZLJLW-UHFFFAOYSA-N
SMILES: NCCP1CC2=CC=C3C=CC=CC3=C2C2=C3C=CC=CC3=CC=C2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results