missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Pyrrolidinyl)phenol, 98%

Catalog No. AAL0817803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0817803 1 g
AAL0817806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0817803 Supplier Thermo Scientific Chemicals Supplier No. L0817803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4787-77-3 | C10H13NO | 163.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 4787-77-3
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00051659
InChI Key ZGSBDRFDXWRZAE-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-yl phenol, 2-pyrrolidinophenol, 2-1-pyrrolidino phenol, 2-1-pyrrolidinyl phenol, 2-tetrahydro-1h-pyrrol-1-ylphenol, 2-pyrrolidinylphenol, 2-pyrrolizinophenol, 2-pyrrolidin-1-yl-phenol, phenol, 2-1-pyrrolidinyl, 1-2-hydroxyphenyl pyrrolidine
PubChem CID 2734845
IUPAC Name 2-pyrrolidin-1-ylphenol
SMILES OC1=CC=CC=C1N1CCCC1

Specifications

Melting Point 109°C to 112°C
Quantity 1 g
Beilstein 135437
Solubility Information Insoluble in water.
Formula Weight 163.22
Percent Purity 98%
Chemical Name or Material 2-(1-Pyrrolidinyl)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only