accessibility menu, dialog, popup

UserName

CAS RN 4787-77-3

IUPAC Name: 2-(pyrrolidin-1-yl)phenol
Synonyms: 2-(pyrrolidin-1-yl)phenol
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: ZGSBDRFDXWRZAE-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results