missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Pyrazolyl)pyridine 98.0+%, TCI America™

Catalog No. P22391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22391G 1 g
P22395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22391G Supplier TCI America Supplier No. P22391G
Only null left

Chemical Identifiers

CAS 25700-11-2
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number MFCD09033534
InChI Key XXTPHXNBKRVYJI-UHFFFAOYSA-N
PubChem CID 5324086
IUPAC Name 2-(1H-pyrazol-1-yl)pyridine
SMILES C1=CN(N=C1)C1=CC=CC=N1

Specifications

Melting Point 39°C
Color White
Quantity 1 g
Formula Weight 145.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-(1-Pyrazolyl)pyridine