missing translation for 'onlineSavingsMsg'
Learn More

[2-(1-Piperidinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™

Catalog No. AC431710500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431710500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431710500 Supplier Thermo Scientific Chemicals Supplier No. 431710500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 480424-81-5 | C12H16BrMgN | 278.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 480424-81-5 , 109-99-9
Molecular Formula C12H16BrMgN
Molecular Weight (g/mol) 278.48
MDL Number MFCD03840894
InChI Key SQFASMRVZVXQCK-UHFFFAOYSA-M
Synonym bromo 2-1-piperidinylmethyl phenyl magnesium, 1-2-bromomagnesio phenyl methyl piperidine, 2-1-piperidinylmethyl phenyl magnesium bromide solution, 2-1-piperidino methyl phenylmagnesium bromide, 2-1-piperidinylmethyl phenyl magnesiu, 2-1-piperidinylmethyl phenyl magnesium bromide solution, 0.25 m in thf, 2-1-piperidino methyl phenylmagnesium bromide 0.25 m in tetrahydrofuran, 2-1-piperidino methyl phenylmagnesium bromide, 0.25m in tetrahydrofuran, grignard reagent
PubChem CID 16217258
IUPAC Name magnesium;1-(phenylmethyl)piperidine;bromide
SMILES Br[Mg]C1=C(CN2CCCCC2)C=CC=C1

Specifications

CAS Min % 4.53, 89.43
CAS Max % 10.57, 95.47
Color Brown
Density 0.9220g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 50 mL
Specific Gravity 0.922
Formula Weight 278.47
Physical Form Liquid
Chemical Name or Material [2-(1-Piperidinylmethyl)phenyl]magnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only