missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperidinyl)aniline, 98%

Catalog No. AAA1307306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1307306 5 g
AAA1307314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1307306 Supplier Thermo Scientific Chemicals Supplier No. A1307306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39643-31-7 | C11H16N2 | 176.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39643-31-7
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00047467
InChI Key OYECAJPUPWFCSL-UHFFFAOYSA-N
Synonym 2-piperidinoaniline, 2-piperidin-1-yl aniline, 2-piperidin-1-yl-phenylamine, benzenamine, 2-1-piperidinyl, n-2-aminophenyl piperidine, 2-1-piperidinyl aniline, 2-1-piperidino aniline, 1-2-aminophenyl piperidine, 2-piperidylphenylamine, 2-piperdinoaniline
PubChem CID 458795
IUPAC Name 2-piperidin-1-ylaniline
SMILES NC1=CC=CC=C1N1CCCCC1

Specifications

Melting Point 44°C to 47°C
Boiling Point 96°C to 98°C (1 mmHg)
Quantity 5 g
UN Number UN2811
Beilstein 151856
Formula Weight 176.26
Percent Purity 98%
Chemical Name or Material 2-(1-Piperidinyl)aniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only